Tyramine hydrochloride
Catalog # EHERC17895800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:173.64
Melting Pt:270…272 °C
EINECS:200-462-5
CAS Number:60-19-5
Formula:C₈H₁₂ClNO
MDL Number:MFCD00012901