Order Entry
Czech Republic
ContactUsLinkComponent
Triethyl orthobenzoate, Sigma-Aldrich®
Triethyl orthobenzoate, Sigma-Aldrich®
Catalog # SIAL75570-50ML
Supplier:  Merck
CAS Number:  
Triethyl orthobenzoate, Sigma-Aldrich®
Catalog # SIAL75570-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :224.3 g/mol
EINECS:216-771-3
CAS Number:1663-61-2
Formula:C₁₃H₂₀O₃
MDL Number:MFCD00009222