
Triethyl orthobenzoate 97%
:1663-61-2
:
:C₁₃H₂₀O₃
:MFCD00009222
:224.3 g/mol
:96 °C
:216-771-3
:GHS07
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4710 to 1.4740 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together