
1,4-Dithioerythritol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6892-68-8
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00063750
:154.25 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:82…83 °C
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:229-998-8
:GHS07
Frequently Bought Together