Ethyl (methylthio)acetate, Sigma-Aldrich®
Catalog # SIAL209716-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:134.2 g/mol
Boiling Pt:70 °C (1013 hPa)
Density:1.04 g/cm³ (20 °C)
ADR:3,III
:M-Clarity™ quality level = MQ100
EINECS:224-700-2
UN:3272
CAS Number:4455-13-4
Formula:C₅H₁₀O₂S
MDL Number:MFCD00009182