

Ethyl (methylthio)acetate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4455-13-4
:
Formula:C₅H₁₀O₂S
MDL Number:MFCD00009182
:134.2 g/mol
Boiling Pt:176 °C
Density:1.06
Flash Pt:59 °C (138 °F)
Storage Temperature:Ambient
EINECS:224-700-2
UN:3272
ADR:3,III
:GHS02
:GHS07
Frequently Bought Together