
2-Hydroxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:582-24-1
:
Formula:C₈H₈O₂
MDL Number:MFCD00041829
:136.15 g/mol
Boiling Pt:118…120 °C (11 mmHg)
Melting Pt:103…105 °C
Density:1.0963 g/cm³ (99 °C)
:M-Clarity™ quality level = MQ100
EINECS:209-480-8
:GHS07
Frequently Bought Together