
6-Phenyl-1-hexanol 97%
Catalog # ACRO308470010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5100 to 1.5120 (20°C, 589 nm)
- GC:≥96.0 %
Specification Test Results
About this item
:
:178.27 g/mol
Boiling Pt:154…155 °C (11 mmHg)
Flash Pt:>110 °C
:GHS07
CAS Number:2430-16-2
Formula:C₁₂H₁₈O
MDL Number:MFCD00014063