D-Glucono-1,5-lactone, Sigma-Aldrich®
: Merck
: GDL, D-Glucono-δ-lactone, D-Gluconic acid-1,5-lactone, delta-Gluconolactone, D-Gluconic acid lactone, (3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-one, D-(+)-Glucono-δ-lactone, D-(+)-Gluconic acid δ-lactone, D-Gluconic acid-δ-lacton, D-(+)-Glucono-1,5-lactone, Gluconolactone
:90-80-2
:
:C₆H₁₀O₆
:MFCD00006647
:178.14 g/mol
:13,04470
:385 °C (1013 hPa)
:153 °C
:1.61 g/cm³ (–5 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:202-016-5
- Environmentally Preferable:
Frequently Bought Together