ContactUsLinkComponent
Dimethyl sulphoxide-[D₆] (99.8% D)
  ABCRAB258209.0050
 :  abcr
 :  
Dimethyl sulphoxide-[D₆] (99.8% D)
  ABCRAB258209.0050
 :  abcr
 :  
  • Pack type:
    Glass bottle
  • Pk:
    50 mL
 :
 :84.09 g/mol
 :189 °C (5 mmHg)
 :20.2 °C
 :1.19 g/cm³ (20 °C)
 :82 °C (closed cup)
 :Ambient
 :218-617-0
 :2206-27-1
 :D₃CS(O)CD₃
 :MFCD00002090