Chloroform-[D₁] 99.8 atom % D + 0.03% (v/v) TMS stabilised for NMR spectroscopy
:865-49-6
:
:CDCl₃
:MFCD00000827
:120.38 g/mol
:60.8 °C (1013 hPa)
:–64 °C
:1.5 g/cm³ (20 °C)
:Refrigerator
:212-742-4
:6.1,III
:Stabilised with silver
:1888
:GHS06
:GHS08
- Appearance:Clear colorless Liquid
- Water:≤0.01 % (Coulometric)
- Stabilizer:silver foil
- D-Enrichment:≥99.80 %
- TMS:0.03 v/v%
Frequently Bought Together