
(1S,2S)-(+)-1,2-Cyclohexanedicarboxylic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:21963-41-7
:
Formula:C₈H₁₂O₄
MDL Number:MFCD00151258
:172.18 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together