(1,1-Dimethoxyethyl)benzene, Sigma-Aldrich®
Catalog # SIAL376760-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:166.22 g/mol
EINECS:224-334-3
CAS Number:4316-35-2
Formula:C₆H₅C(OCH₃)₂CH₃
MDL Number:MFCD00075617