Pour spout, Nalgene®
38 - 430
NALGDS2176-0384
PP, translucent. with polyethylene insert.
- Used for all Nalgene bottles with 38-430 neck finish
- Good chemical resistance
- Good temperature resistance
Allows easy one-handed liquid dispensing without leaving bottles completely open to the environment.
- Thread:38 - 430
- Cat. No.:NALGDS2176-0384
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact