Order Entry
Czech Republic
ContactUsLinkComponent
Acetonitrile, BAKER BIO-ANALYZED®, J.T.Baker®
Acetonitrile, BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # 8144.4000GL
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  
undefined
Acetonitrile, BAKER BIO-ANALYZED®, J.T.Baker®
Catalog # 8144.4000GL
Supplier:  Avantor
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Specifications

Specification Test Results

  • Assay (corrected for H2O):
    min. 99.9 %
  • Residue after Evaporation:
    max. 1 ppm
  • Titrable Acid (µeq/g):
    max. 0.5
  • Titrable base (µeq/g):
    max. 0.6
  • Water (H2O):
    max. 30 ppm
  • Measured at 450 nm:
    max. 0.3
  • Measured at Emission Maximum for Solvent Impurities:
    max. 1.0
  • at 200 nm:
    max. 0.05
  • at 220 nm:
    max. 0.01
  • at 254-400 nm:
    max. 0.01
  • UV Cut-off, nm:
    max. 190

Specification Test Results

About this item

 :
 :41.05 g/mol
Merck Index:13,00071
Boiling Pt:81.6 °C (1013 hPa)
Melting Pt:–45.7 °C
Density:0.782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
 :GHS02
 :GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878