

Acetonitrile ≥99.8% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
Merck Index:14,70
Boiling Pt:80…82 °C
Melting Pt:–48 °C…–45 °C
Density:0.786
Flash Pt:5 °C (41 °F)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
REACH:01-2119471307-38
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Related Information
Frequently Bought Together