Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
Supplier:   Brand
Desiccator plates
Supplier:  Brand
Certificates
Desiccator plates
Desiccator plates
6 Options Available Below
Product Details & Documents
Desiccator plates for glass and plastic desiccators.
  • Central hole measures approximately 20 mm diameter
  • Small perforations measure approximately 5 mm diameter
Product Family Options
Porcelain
Product Information
  • Type
    Ø
    Availability
    Price
  • Porcelain
    140 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 1 790,00 Kč
    1 790,00 Kč
    1 * 1 KS
       1 790,00 Kč
  • Porcelain
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 3 490,00 Kč
    3 490,00 Kč
    1 * 1 KS
       3 490,00 Kč
  • Porcelain
    235 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 4 150,00 Kč
    4 150,00 Kč
    1 * 1 KS
       4 150,00 Kč
PP
Product Information
  • Type
    Ø
    Availability
    Price
  • PP
    140 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 390,00 Kč
    390,00 Kč
    1 * 1 KS
       390,00 Kč
  • PP
    190 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 515,00 Kč
    515,00 Kč
    1 * 1 KS
       515,00 Kč
  • PP
    235 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 535,00 Kč
    535,00 Kč
    1 * 1 KS
       535,00 Kč