1,3-Dibromoacetone, Technical Grade
Catalog # ABCRAB174501.0005
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:217 °C (1013 hPa)
Melting Pt:28 °C
Density:2.167 g/cm³ (18 °C)
ADR:8,II
:215.87 g/mol
EINECS:212-430-8
UN:1760
CAS Number:816-39-7
Formula:BrH₂CC(O)CH₂Br
MDL Number:MFCD00013540