

2-Bromopropane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-26-3
:
Formula:H₃CBrCHCH₃
MDL Number:MFCD00000147
Merck Index:13,05230
Boiling Pt:59 °C (760 mmHg)
Melting Pt:–89 °C
Density:1.31 g/cm³
Flash Pt:1 °C
UN:2344
ADR:3,II
:122.99 g/mol
EINECS:200-855-1
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4250 to 1.4270 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together