Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
2-Bromobutane 99+%
 
undefined
 
 
2-Bromobutane 99+%
 

 

  • Appearance:
    Clear colorless to yellow-brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4359 to 1.4379 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Titration with HClO4:
    ≥98.5 %
  • Sensitivity:
    (Test for Ni) Passes

 

 

Frequently Bought Together