Order Entry
Czech Republic
ContactUsLinkComponent
 
Environment
1-Bromodecane for synthesis, Sigma-Aldrich®
Catalog # 8.01676.0250
Supplier:  Merck
CAS Number:  
Environment
1-Bromodecane for synthesis, Sigma-Aldrich®
Catalog # 8.01676.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -30 °C
  • Molar Mass:
    221.17 g/mol
  • Boiling Point:
    238 °C (1013 mbar)
  • Flash Point:
    94 °C
  • Refractive Index:
    1.456 (20 °C, 589 nm)
  • Density:
    1.07 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :221.18 g/mol
Boiling Pt:238 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.07 g/cm³ (20 °C)
Flash Pt:94 °C
Storage Temperature:Ambient
EINECS:203-955-3
UN:3082
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS09
CAS Number:112-29-8
Formula:BrH₂C(CH₂)₈CH₃
MDL Number:MFCD00000221