1,4-Dibromobutane ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,4-Dibromo butane
CAS Number:110-52-1
:
Formula:BrH₂C(CH₂)₂CH₂Br
MDL Number:MFCD00000261
:215.92 g/mol
Boiling Pt:194…196 °C
Melting Pt:–20 °C
Density:1.820
Flash Pt:131 °C (267 °F)
Storage Temperature:Ambient
EINECS:203-775-5
UN:2810
ADR:6.1,III
:GHS06
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together