


1,2-Dibromoethane 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-93-4
:
Formula:BrH₂CCH₂Br
MDL Number:MFCD00000233
:187.86 g/mol
Merck Index:13,03830
Boiling Pt:131…132°C (760mmHg)
Melting Pt:9…10°C
Density:2.173
Flash Pt:>104°C
Storage Temperature:Ambient
EINECS:203-444-5
UN:1605
ADR:6.1,I
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:clear colorless to light yellow liquid
- Infrared spectrum:Conforms
- GC:≥98.5%
- Refractive index:1.5375 to 1.5395 (20°C, 589nm)
Specification Test Results
Frequently Bought Together