Order Entry
Czech Republic
ContactUsLinkComponent
Silica gel drying agent, with moisture indicator (orange gel), Supelco®
Silica gel drying agent, with moisture indicator (orange gel), Supelco®
Catalog # 1.03806.0001
Supplier:  Merck
CAS Number:  
Silica gel drying agent, with moisture indicator (orange gel), Supelco®
Catalog # 1.03806.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Metal can
  • Pk:
    10
  • Seq:
    0010

Specifications

Specification Test Results

  • Solubility:
    1 g/l
  • Melting Point:
    1000 °C
  • Water Absorption:
    250 g/kg (20 °C)

Specification Test Results

About this item

 :
 :60.08 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232