Order Entry
Czech Republic
ContactUsLinkComponent
Lanthanum(III) nitrate hexahydrate ≥99%, GPR RECTAPUR®
Oxidizing
Irritant
Lanthanum(III) nitrate hexahydrate ≥99%, GPR RECTAPUR®
Catalog # 24958.238
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oxidizing
Irritant
Lanthanum(III) nitrate hexahydrate ≥99%, GPR RECTAPUR®
Catalog # 24958.238
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (calculated as La2O3):
    Min. 99 %

Specification Test Results

About this item

 :
 :433.01 g/mol
Merck Index:14,05363
Boiling Pt:126 °C (1013 hPa)
Melting Pt:65…67 °C
Density:2.347 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-238-0
UN:1477
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:10277-43-7
 :10227-43-7
Formula:La(NO₃)₃·6H₂O
MDL Number:MFCD00149751