


Lanthanum(III) nitrate hexahydrate analytical reagent, Supelco®
Catalog # 1.05326.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:1580 g/l (25 °C)
- Melting Point:40 °C
- Molar Mass:433.02 g/mol
- Bulk Density:1300 kg/m3
- Boiling Point:126 °C (1013 hPa) (decomposition)
- pH:3.4 (50 g/l, H2O, 25 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
About this item
:
:433.01 g/mol
Merck Index:14,05363
Boiling Pt:126 °C (1013 hPa)
Melting Pt:65…67 °C
Density:2.347 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-238-0
UN:1477
ADR:5.1,II
:M-Clarity™ quality level = MQ100
:GHS03
:GHS05
:GHS09
CAS Number:10277-43-7
:10227-43-7
Formula:La(NO₃)₃·6H₂O
MDL Number:MFCD00149751