

Lanthanum(III) nitrate hexahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10277-43-7
:10227-43-7
Formula:La(NO₃)₃·6H₂O
MDL Number:MFCD00149751
:433.01 g/mol
Merck Index:14,5363
Boiling Pt:126 °C (1013 hPa)
Melting Pt:40 °C
Density:2.347 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-238-0
UN:1477
ADR:5.1,III
:GHS03
:GHS07
Related Information
Frequently Bought Together