Order Entry
Czech Republic
ContactUsLinkComponent
Tin(IV) chloride pentahydrate 98+%, extra pure
Corrosive
Irritant
Tin(IV) chloride pentahydrate 98+%, extra pure
Certificates
undefined
Corrosive
Irritant
Tin(IV) chloride pentahydrate 98+%, extra pure

Specification Test Results

  • Appearance:
    White to off-white Powder or crystals and/or lumps
  • Assay:
    ≥98.0 %
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm

Specification Test Results

Related Information

Frequently Bought Together