Order Entry
Czech Republic
ContactUsLinkComponent
Tin(II) chloride dihydrate, EMSURE® for analysis (max. 0.01 ppm Hg), Supelco®
Corrosive
Health
Irritant
Environment
Tin(II) chloride dihydrate, EMSURE® for analysis (max. 0.01 ppm Hg), Supelco®
Catalog # 1.07814.2500
Supplier:  Merck
CAS Number:  
Corrosive
Health
Irritant
Environment
Tin(II) chloride dihydrate, EMSURE® for analysis (max. 0.01 ppm Hg), Supelco®
Catalog # 1.07814.2500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Solubility:
    1187 g/l (20 °C)
  • Melting Point:
    246 °C (anhydrous substance)
  • Molar Mass:
    225.63 g/mol
  • Bulk Density:
    1250 kg/m3
  • Boiling Point:
    623 °C (1013 hPa) (anhydrous substance)
  • Density:
    2.71 g/cm3 (20 °C)
  • pH:
    1 - 2 (100 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :225.65 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:2.71 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-868-0
UN:3260
ADR:8,II
 :M-Clarity™ quality level = MQ300
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:10025-69-1
Formula:SnCl₂·2H₂O
MDL Number:MFCD00149863