Order Entry
Czech Republic
ContactUsLinkComponent
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Not precipitated by NH4OH (as SO4):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

Frequently Bought Together