Aluminium nitrate nonahydrate, Supelco®
Catalog # 1.01086.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:375.13 g/mol
Melting Pt:73 °C
Density:1.72 g/cm³ (20 °C)
ADR:5.1,III
:M-Clarity™ quality level = MQ200
EINECS:236-751-8
UN:1438
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7784-27-2
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00149132