Aluminium nitrate nonahydrate, EMSURE® for analysis, Supelco®
1.01063.0500
: Merck
:
- Pack type:Plastic bottle
- Pk:500 g
- Seq:3645
- Solubility:419 g/l (20 °C)
- Melting Point:73 °C
- Molar Mass:375.13 g/mol
- Bulk Density:880 kg/m3
- pH:2.0 - 4.0 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
:
:375.13 g/mol
:73 °C
:1.72 g/cm³ (20 °C)
:5.1,III
:M-Clarity™ quality level = MQ300
:236-751-8
:1438
:GHS05
:GHS07
:Ambient
:7784-27-2
:Al(NO₃)₃·9H₂O
:MFCD00149132