


1-Iodo-2-methylpropane ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:513-38-2
:
Formula:C₄H₉I
MDL Number:MFCD00001084
:184.02 g/mol
Merck Index:14,5144
Boiling Pt:120…121 °C
Melting Pt:–93 °C
Density:1.599
Flash Pt:12 °C (53 °F)
Storage Temperature:Ambient
UN:2391
ADR:3,II
:Stabilised with copper
EINECS:208-160-5
:GHS02
:GHS06
:GHS07
Related Information
Frequently Bought Together