
2-Ethylhexyl bromide 96% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18908-66-2
:
Formula:C₈H₁₇Br
MDL Number:MFCD00000220
:193.13 g/mol
Boiling Pt:94…95 °C (35 mmHg)
Density:1.086
Flash Pt:69 °C (156 °F)
:Stabilised with copper
EINECS:242-659-9
:GHS07
Storage Temperature:Refrigerator
Related Information
Certificates
Search by Lot NumberOrganic Building Blocks and Intermediates
Application Notes
Thermo Scientific Organic Photovoltaics
Product Literature
Frequently Bought Together