Order Entry
Czech Republic
ContactUsLinkComponent
Calcium nitrate tetrahydrate ≥98%, GPR RECTAPUR®
Calcium nitrate tetrahydrate ≥98%, GPR RECTAPUR®
Catalog # 22399.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium nitrate tetrahydrate ≥98%, GPR RECTAPUR®
Catalog # 22399.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 0.02 %
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
 :GHS07
 :GHS05
CAS Number:13477-34-4
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604