


Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-88-4
:
Formula:CH₃I
MDL Number:MFCD00001073
:141.94 g/mol
Merck Index:14,6087
Boiling Pt:41…43 °C
Melting Pt:–67 °C
Density:2.28
Storage Temperature:Refrigerator
UN:2644
ADR:6.1,I
:Stabilised with copper
EINECS:200-819-5
:GHS06
:GHS08
:GHS07
Related Information
Frequently Bought Together