Order Entry
Czech Republic
ContactUsLinkComponent
Calcium nitrate tetrahydrate, Supelco®
Calcium nitrate tetrahydrate, Supelco®
Catalog # 1.02120.5000
Supplier:  Merck
CAS Number:  
Calcium nitrate tetrahydrate, Supelco®
Catalog # 1.02120.5000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

About this item

 :
 :236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS07
CAS Number:13477-34-4
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604