ContactUsLinkComponent
1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilised
  A10954.06
 :  
1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilised
  A10954.06
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
 :
 :353.82 g/mol
 :112…113 °C
 :–21 °C
 :2.629
 :Stabilised with copper
 :Ambient
 :206-567-2
 :GHS07
 :354-65-4
 :C₂F₄I₂
 :MFCD00039401