
Strontium chloride hexahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-70-4
:
Formula:SrCl₂·6H₂O
MDL Number:MFCD00149865
:266.62 g/mol
Merck Index:14,8840
Boiling Pt:100 °C (1013 hPa)
Melting Pt:61 °C
Density:1.93
EINECS:233-971-6
:GHS05
Storage Temperature:Ambient
Related Information
Frequently Bought Together