Order Entry
Czech Republic
ContactUsLinkComponent
Magnesium chloride hexahydrate, Sigma-Aldrich®
Magnesium chloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL63033-1KG-F
Supplier:  Merck
CAS Number:  
Magnesium chloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL63033-1KG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :203.3 g/mol
Merck Index:13,05684
Boiling Pt:1412 °C (1013 hPa)
Melting Pt:117 °C
Density:1.57 g/cm³ (25 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:232-094-6
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781