
Barium chloride dihydrate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10326-27-9
:
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154
:244.27 g/mol
Merck Index:14,00971
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:960 °C
Density:3.86 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:GHS06
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 20 ppm
- Insolubility in water:Max. 0.05 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 750 ml Plastic bottle for solids DIN 60 (ZPACK13250750)
Packaging Information
spec_VWRC21715.263 spec_VWRC21715.263 spec_VWRC21715.296 spec_VWRC21715.296
Product specification
Frequently Bought Together