
Barium chloride dihydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:10326-27-9
:
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154
:244.27 g/mol
Merck Index:14,00971
Boiling Pt:1560 °C (1013 hPa)
Melting Pt:960 °C
Density:3.86 g/cm³ (25 °C)
UN:1564
ADR:6.1,III
:M-Clarity™ quality level = MQ300
EINECS:233-788-1
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Solubility:358 g/l (20 °C) (anhydrous substance)
- Melting Point:963 °C (anhydrous substance)
- Molar Mass:244.28 g/mol
- Bulk Density:1200 - 1400 kg/m3
- Boiling Point:1560 °C (anhydrous substance)
- Density:3.86 g/cm3
- pH:5.2 - 8.0 (50 g/l, H2O, 25 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.01719.0500_TDS_EN ProductPage_101719
Product specification
msds_101719
SDS
Frequently Bought Together