
Barium chloride dihydrate ≥99%, Puriss. p.a. plus ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:10326-27-9
:
Formula:BaCl₂·2H₂O
MDL Number:MFCD00149154
:244.27 g/mol
Merck Index:14,00971
Boiling Pt:1.560 °C (1.013 hPa)
Melting Pt:962 °C
Density:3.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:GHS06
Specification Test Results
- Assay:Min. 99.0 %
- Loss on drying (150°C):14.0 - 16.0 %
- Water insoluble matter:Max. 0.005 %
- pH (5 %, 25 °C):5.2 - 8.0
- Calcium (Ca):Max. 0.005 %
- Iron (Fe):Max. 1 ppm
- Potassium (K):Max. 0.0025 %
- Magnesium (Mg):Max. 0.001 %
- Sodium (Na):Max. 0.005 %
- Lead (Pb):Max. 0.001 %
- Strontium (Sr):Max. 0.01 %
- Heavy metals (ICP) ACS:Max. 5 ppm
- Oxidizing substances (as NO₃):Max. 0.005 %
- Total N:Max. 0.002 %
Specification Test Results
Frequently Bought Together