
Sodium tungstate dihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10213-10-2
:
Formula:Na₂WO₄·2H₂O
MDL Number:MFCD00149190
:329.85 g/mol
Merck Index:12,08847
Melting Pt:100 °C
Density:3.23 g/cm³ (20 °C)
EINECS:236-743-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together