Order Entry
Czech Republic
ContactUsLinkComponent
Sodium tungstate dihydrate, Supelco®
Irritant
Sodium tungstate dihydrate, Supelco®
Catalog # 1.06672.1000
Supplier:  Merck
CAS Number:  
undefined
Irritant
Sodium tungstate dihydrate, Supelco®
Catalog # 1.06672.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :329.85 g/mol
Merck Index:12,08847
Melting Pt:100 °C
Density:3.23 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:236-743-4
 :GHS07
Storage Temperature:Ambient
CAS Number:10213-10-2
Formula:Na₂WO₄·2H₂O
MDL Number:MFCD00149190