Order Entry
Czech Republic
ContactUsLinkComponent
Lithium sulphate monohydrate, Suprapur® for trace analysis, Supelco®
Irritant
Lithium sulphate monohydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.05697.0050
Supplier:  Merck
CAS Number:  
Irritant
Lithium sulphate monohydrate, Suprapur® for trace analysis, Supelco®
Catalog # 1.05697.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    340 g/l (20 °C)
  • Melting Point:
    120 °C
  • Molar Mass:
    127.96 g/mol
  • Bulk Density:
    940 kg/m3
  • Density:
    2.05 g/cm3 (20 °C)
  • pH:
    4.5 - 6.0 (50 g/l, H2O, 20 °C)

Specification Test Results

About this item

 :
 :127.96 g/mol
Melting Pt:120 °C
Density:2.05 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:233-820-4
 :GHS07
Storage Temperature:Ambient
CAS Number:10102-25-7
Formula:Li₂SO₄·H₂O
MDL Number:MFCD00149766