Order Entry
Czech Republic
ContactUsLinkComponent
Sodium thiosulphate pentahydrate ≥99%, Proteomics Grade
Sodium thiosulphate pentahydrate ≥99%, Proteomics Grade
Catalog # M132-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium thiosulphate pentahydrate ≥99%, Proteomics Grade
Catalog # M132-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Insolubles:
    <0.005%
  • pH (5%, Water) @25 °C:
    6 – 8.4
  • Protease:
    none detected
  • Purity:
    >99%
  • Sulphate and Sulphite:
    <0.1%
  • Sulphide:
    <0.0001%

Specification Test Results

About this item

 :
 :248.19 g/mol
Merck Index:12,08844
Melting Pt:49 °C
Density:1.67 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-867-5
ADR:,
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186