Disodium hydrogen phosphate dihydrate, Sigma-Aldrich®
Catalog # SIAL71662-1KG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:177.99 g/mol
Melting Pt:92.5 °C
Density:2.1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-448-7
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182