Order Entry
Czech Republic
ContactUsLinkComponent
Tetrasodium diphosphate decahydrate 99+% for analysis
Irritant
Tetrasodium diphosphate decahydrate 99+% for analysis
Certificates
undefined
Irritant
Tetrasodium diphosphate decahydrate 99+% for analysis

Specification Test Results

  • Appearance:
    White Crystals
  • Titration with HClO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤10 ppm
  • Iron (Fe):
    ≤5 ppm
  • Potassium (K):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤5 ppm
  • Total nitrogen compounds:
    ≤0.001 % (as N)
  • Impurity:
    ≤0.1 % (ortho-phosphate (as PO4))

Specification Test Results

Related Information

Frequently Bought Together