Order Entry
Czech Republic
ContactUsLinkComponent
 
 
 
 
Sodium perchlorate monohydrate ≥99.0% for HPLC, Fluka™
 
 
 
 
Sodium perchlorate monohydrate ≥99.0% for HPLC, Fluka™

 

  • Assay:
    99.0 - 101.0 %
  • Solution 5% in H₂O:
    complying
  • pH (5 %, 25 °C):
    5.0 - 8.0
  • Absorbance at 210 nm:
    Max. 0.1
  • Absorbance at 220 nm:
    Max. 0.05
  • Absorbance at 230 nm:
    Max. 0.03
  • Absorbance at 240 nm:
    Max. 0.02
  • Absorbance 500 nm:
    Max. 0.02

 

 

Frequently Bought Together